Assay Details
| Gene Symbol: Klf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845254.3 | XP_004845311.1 | 3 - 4 | 1491 | 102 | |
| XM_004845255.3 | XP_004845312.1 | 3 - 4 | 1111 | 102 | ||
More Information
Set Membership: |
|

Klf5
Genomic Map