Assay Details
| Gene Symbol: LOC101712008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845721.2 | XP_004845778.1 | 5 - 6 | 1214 | 83 | |
| XM_004845722.2 | XP_004845779.1 | 4 - 5 | 1151 | 83 | ||
More Information
Set Membership: |
|

LOC101712008
Genomic Map