Assay Details
| Gene Symbol: LOC101723225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848054.3 | XP_004848111.1 | 15 - 16 | 2019 | 100 | |
| XM_021261057.1 | XP_021116716.1 | 15 - 16 | 2369 | 100 | ||
More Information
Set Membership: |
|

LOC101723225
Genomic Map