Assay Details
| Gene Symbol: Brd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848996.3 | XP_004849053.1 | 9 - 10 | 1877 | 65 | |
| XM_013073779.2 | XP_012929233.1 | 9 - 10 | 1915 | 65 | ||
More Information
Set Membership: |
|

Brd3
Genomic Map