Assay Details
| Gene Symbol: Otud3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850454.2 | XP_004850511.1 | 6 - 7 | 993 | 64 | |
| XM_021262956.1 | XP_021118615.1 | 6 - 7 | 766 | 64 | ||
More Information
Set Membership: |
|

Otud3
Genomic Map