Assay Details
| Gene Symbol: Ncmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850596.2 | XP_004850653.1 | 3 - 4 | 360 | 84 | |
| XM_021263073.1 | XP_021118732.1 | 4 - 5 | 439 | 84 | ||
| XM_021263074.1 | XP_021118733.1 | 2 - 3 | 396 | 84 | ||
More Information
Set Membership: |
|

Ncmap
Genomic Map