Assay Details
| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853542.2 | XP_004853599.1 | 10 - 11 | 1132 | 71 | |
| XM_013075354.2 | XP_012930808.1 | 10 - 11 | 1170 | 71 | ||
| XM_021265557.1 | XP_021121216.1 | 10 - 11 | 1018 | 71 | ||
More Information
Set Membership: |
|

Ppp2r5a
Genomic Map