Assay Details
| Gene Symbol: Aspscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860995.3 | XP_004861052.1 | 13 - 14 | 1499 | 61 | |
| XM_004860996.3 | XP_004861053.1 | 13 - 14 | 1511 | 61 | ||
More Information
Set Membership: |
|

Aspscr1
Genomic Map