Assay Details
| Gene Symbol: CUNH14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864186.2 | XP_004864243.1 | 2 - 3 | 1066 | 105 | |
| XM_021243880.1 | XP_021099539.1 | 5 - 6 | 2086 | 105 | ||
| XM_021243881.1 | XP_021099540.1 | 4 - 5 | 2007 | 105 | ||
| XM_021243882.1 | XP_021099541.1 | 5 - 6 | 2104 | 105 | ||
| XM_021243883.1 | XP_021099542.1 | 5 - 6 | 2118 | 105 | ||
More Information
Set Membership: |
|

CUNH14orf93
Genomic Map