Assay Details
| Gene Symbol: CUNH14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864186.2 | XP_004864243.1 | 3 - 4 | 1385 | 87 | |
| XM_004864187.2 | XP_004864244.1 | 3 - 4 | 1316 | 87 | ||
| XM_004864188.3 | XP_004864245.1 | 2 - 3 | 1451 | 87 | ||
| XM_021243880.1 | XP_021099539.1 | 6 - 7 | 2405 | 87 | ||
| XM_021243881.1 | XP_021099540.1 | 5 - 6 | 2326 | 87 | ||
| XM_021243882.1 | XP_021099541.1 | 6 - 7 | 2423 | 87 | ||
| XM_021243883.1 | XP_021099542.1 | 6 - 7 | 2437 | 87 | ||

CUNH14orf93
Genomic Map