Assay Details
| Gene Symbol: CUNH14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864186.2 | XP_004864243.1 | 5 - 6 | 1554 | 74 | |
| XM_004864187.2 | XP_004864244.1 | 5 - 6 | 1485 | 74 | ||
| XM_004864188.3 | XP_004864245.1 | 4 - 5 | 1620 | 74 | ||
| XM_021243880.1 | XP_021099539.1 | 8 - 9 | 2574 | 74 | ||
| XM_021243881.1 | XP_021099540.1 | 7 - 8 | 2495 | 74 | ||
| XM_021243882.1 | XP_021099541.1 | 8 - 9 | 2592 | 74 | ||
| XM_021243883.1 | XP_021099542.1 | 8 - 9 | 2606 | 74 | ||
More Information
Set Membership: |
|

CUNH14orf93
Genomic Map