Assay Details
| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866217.3 | XP_004866274.1 | 9 - 10 | 1703 | 89 | |
| XM_004866218.3 | XP_004866275.1 | 9 - 10 | 1701 | 89 | ||
| XM_004866219.3 | XP_004866276.1 | 8 - 9 | 1618 | 89 | ||
| XM_004866220.3 | XP_004866277.1 | 8 - 9 | 1616 | 89 | ||
| XM_013066077.2 | XP_012921531.1 | 9 - 10 | 1435 | 89 | ||
| XM_013066078.2 | XP_012921532.1 | 9 - 10 | 1432 | 89 | ||
| XM_021245457.1 | XP_021101116.1 | 10 - 11 | 1712 | 89 | ||
| XM_021245458.1 | XP_021101117.1 | 10 - 11 | 1440 | 89 | ||
| XM_021245460.1 | XP_021101119.1 | 7 - 8 | 1514 | 89 | ||
| XM_021245461.1 | XP_021101120.1 | 8 - 9 | 1304 | 89 | ||
More Information
Set Membership: |
|

Lef1
Genomic Map