Assay Details
| Gene Symbol: LOC101712254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868633.3 | XP_004868690.1 | 1 - 2 | 145 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004868633.3 transcripts.
Set Membership: |
|

LOC101712254
Genomic Map