Assay Details
| Gene Symbol: Fggy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869604.3 | XP_004869661.1 | 12 - 13 | 1434 | 64 | |
| XM_004869605.3 | XP_004869662.1 | 12 - 13 | 1422 | 64 | ||
| XM_004869606.2 | XP_004869663.1 | 12 - 13 | 1366 | 64 | ||
| XM_021248220.1 | XP_021103879.1 | 11 - 12 | 1326 | 64 | ||
| XM_021248222.1 | XP_021103881.1 | 11 - 12 | 1126 | 64 | ||
| XM_021248223.1 | XP_021103882.1 | 9 - 10 | 1099 | 64 | ||
| XM_021248224.1 | XP_021103883.1 | 9 - 10 | 1161 | 64 | ||
| XM_021248225.1 | XP_021103884.1 | 7 - 8 | 928 | 64 | ||
More Information
Set Membership: |
|

Fggy
Genomic Map