Assay Details
| Gene Symbol: Fdps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871133.3 | XP_004871190.1 | 5 - 6 | 1086 | 52 | |
| XM_004871134.3 | XP_004871191.1 | 5 - 6 | 696 | 52 | ||
| XM_004871135.3 | XP_004871192.1 | 5 - 6 | 673 | 52 | ||
| XM_021249255.1 | XP_021104914.1 | 5 - 6 | 1065 | 52 | ||
More Information
Set Membership: |
|

Fdps
Genomic Map