Assay Details
Gene Symbol: LOC101723821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013073804.2 | XP_012929258.1 | 10 - 11 | 1641 | 95 | |
XM_013073805.2 | XP_012929259.1 | 10 - 11 | 1647 | 95 | ||
XM_013073806.2 | XP_012929260.1 | 9 - 10 | 1456 | 95 | ||
XM_013073807.2 | XP_012929261.1 | 8 - 9 | 1258 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
