Assay Details
| Gene Symbol: LOC101723821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073804.2 | XP_012929258.1 | 12 - 13 | 1921 | 88 | |
| XM_013073806.2 | XP_012929260.1 | 11 - 12 | 1736 | 88 | ||
| XM_013073807.2 | XP_012929261.1 | 10 - 11 | 1538 | 88 | ||
More Information
Set Membership: |
|

LOC101723821
Genomic Map