Assay Details
| Gene Symbol: LOC101723821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073804.2 | XP_012929258.1 | 17 - 19 | 2469 | 117 | |
| XM_013073805.2 | XP_012929259.1 | 16 - 18 | 2412 | 117 | ||
| XM_013073806.2 | XP_012929260.1 | 16 - 18 | 2284 | 117 | ||
| XM_013073807.2 | XP_012929261.1 | 15 - 17 | 2086 | 117 | ||
More Information
Set Membership: |
|

LOC101723821
Genomic Map