Assay Details
| Gene Symbol: LOC101723821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073804.2 | XP_012929258.1 | 8 - 9 | 1226 | 84 | |
| XM_013073805.2 | XP_012929259.1 | 8 - 9 | 1232 | 84 | ||
More Information
Set Membership: |
|

LOC101723821
Genomic Map