Assay Details
| Gene Symbol: Ceacam16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250894.1 | XP_021106553.1 | 5 - 6 | 1104 | 55 | |
| XM_021250895.1 | XP_021106554.1 | 5 - 6 | 1104 | 55 | ||
| XM_021250896.1 | XP_021106555.1 | 7 - 8 | 2573 | 55 | ||
| XM_021250897.1 | XP_021106556.1 | 7 - 8 | 2498 | 55 | ||
| XM_021250898.1 | XP_021106557.1 | 5 - 6 | 1104 | 55 | ||
| XM_021250900.1 | XP_021106559.1 | 5 - 6 | 1008 | 55 | ||
| XM_021250901.1 | XP_021106560.1 | 4 - 5 | 987 | 55 | ||
| XM_021250902.1 | XP_021106561.1 | 6 - 7 | 984 | 55 | ||
| XM_021250903.1 | XP_021106562.1 | 6 - 7 | 2451 | 55 | ||
| XM_021250904.1 | XP_021106563.1 | 5 - 6 | 931 | 55 | ||
More Information
Set Membership: |
|

Ceacam16
Genomic Map