Assay Details
| Gene Symbol: Txndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260680.1 | XP_021116339.1 | 7 - 8 | 730 | 56 | |
| XM_021260681.1 | XP_021116340.1 | 8 - 9 | 840 | 56 | ||
More Information
Set Membership: |
|

Txndc5
Genomic Map