Assay Details
| Gene Symbol: Fzd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262752.1 | XP_021118411.1 | 3 - 4 | 542 | 63 | |
| XM_021262753.1 | XP_021118412.1 | 3 - 4 | 558 | 63 | ||
More Information
Set Membership: |
|

Fzd6
Genomic Map