Assay Details
| Gene Symbol: Otud7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021264727.1 | XP_021120386.1 | 4 - 5 | 556 | 64 | |
| XM_021264728.1 | XP_021120387.1 | 5 - 6 | 685 | 64 | ||
| XM_021264729.1 | XP_021120388.1 | 4 - 5 | 556 | 64 | ||
| XM_021264730.1 | XP_021120389.1 | 5 - 6 | 597 | 64 | ||
| XM_021264731.1 | XP_021120390.1 | 4 - 5 | 593 | 64 | ||
More Information
Set Membership: |
|

Otud7a
Genomic Map