Assay Details
| Gene Symbol: Kifc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846801.3 | XP_004846858.2 | 7 - 8 | 1679 | 57 | |
| XM_013073001.2 | XP_012928455.1 | 7 - 8 | 1812 | 57 | ||
| XM_013073002.2 | XP_012928456.1 | 7 - 8 | 1757 | 57 | ||
| XM_021260020.1 | XP_021115679.1 | 6 - 7 | 1636 | 57 | ||
| XM_021260021.1 | XP_021115680.1 | 7 - 8 | 1881 | 57 | ||
| XM_021260022.1 | XP_021115681.1 | 7 - 8 | 1961 | 57 | ||
More Information
Set Membership: |
|

Kifc1
Genomic Map