Assay Details
| Gene Symbol: Cda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850469.2 | XP_004850526.1 | 1 - 2 | 250 | 71 | |
| XM_004850470.2 | XP_004850527.1 | 1 - 2 | 240 | 71 | ||
More Information
Set Membership: |
|

Cda
Genomic Map