Assay Details
| Gene Symbol: Zbtb8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850864.2 | XP_004850921.1 | 4 - 5 | 1473 | 84 | |
| XM_004850865.2 | XP_004850922.1 | 5 - 6 | 1325 | 84 | ||
| XM_004850867.2 | XP_004850924.1 | 4 - 5 | 1281 | 84 | ||
| XM_013074248.2 | XP_012929702.1 | 4 - 5 | 1167 | 84 | ||
More Information
Set Membership: |
|

Zbtb8a
Genomic Map