Assay Details
Gene Symbol: CUNH11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852191.2 | XP_004852248.1 | 3 - 4 | 511 | 90 | |
XM_021264482.1 | XP_021120141.1 | 2 - 3 | 308 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
