Assay Details
| Gene Symbol: Lipn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858350.2 | XP_004858407.1 | 5 - 6 | 689 | 65 | |
| XM_021239174.1 | XP_021094833.1 | 6 - 7 | 830 | 65 | ||
| XM_021239175.1 | XP_021094834.1 | 6 - 7 | 806 | 65 | ||
| XM_021239176.1 | XP_021094835.1 | 6 - 7 | 689 | 65 | ||
| XM_021239177.1 | XP_021094836.1 | 6 - 7 | 748 | 65 | ||
More Information
Set Membership: |
|

Lipn
Genomic Map