Assay Details
| Gene Symbol: St8sia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859967.2 | XP_004860024.1 | 2 - 3 | 234 | 63 | |
| XM_021240480.1 | XP_021096139.1 | 4 - 5 | 533 | 63 | ||
More Information
Set Membership: |
|

St8sia6
Genomic Map