Assay Details
| Gene Symbol: Znf775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860416.3 | XP_004860473.1 | 1 - 2 | 110 | 103 | |
| XM_013078060.2 | XP_012933514.1 | 1 - 2 | 110 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004860416.3, XM_013078060.2 transcripts.
Set Membership: |
|

Znf775
Genomic Map