Assay Details
| Gene Symbol: Znf775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860416.3 | XP_004860473.1 | 2 - 3 | 190 | 90 | |
| XM_004860418.3 | XP_004860475.1 | 2 - 3 | 1657 | 90 | ||
| XM_021240811.1 | XP_021096470.1 | 3 - 4 | 1612 | 90 | ||
| XM_021240812.1 | XP_021096471.1 | 3 - 4 | 1456 | 90 | ||
| XM_021240813.1 | XP_021096472.1 | 3 - 4 | 1576 | 90 | ||
| XM_021240814.1 | XP_021096473.1 | 2 - 3 | 294 | 90 | ||
| XM_021240815.1 | XP_021096474.1 | 2 - 3 | 258 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004860416.3, XM_004860418.3, XM_021240811.1, XM_021240812.1, XM_021240813.1, XM_021240814.1, XM_021240815.1 transcripts.
Set Membership: |
|

Znf775
Genomic Map