Assay Details
| Gene Symbol: Pou3f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862495.2 | XP_004862552.1 | 1 | 222 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004862495.2 transcripts.
Set Membership: |
|

Pou3f2
Genomic Map