Assay Details
| Gene Symbol: Il17rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864333.2 | XP_004864390.1 | 1 - 2 | 137 | 57 | |
| XM_013065450.2 | XP_012920904.1 | 1 - 2 | 137 | 57 | ||
More Information
Set Membership: |
|

Il17rb
Genomic Map