Assay Details
| Gene Symbol: Gna13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870274.3 | XP_004870331.1 | 2 - 3 | 609 | 79 | |
| XM_021248584.1 | XP_021104243.1 | 2 - 3 | 610 | 79 | ||
| XM_021248585.1 | XP_021104244.1 | 3 - 4 | 400 | 79 | ||
More Information
Set Membership: |
|

Gna13
Genomic Map