Assay Details
| Gene Symbol: LOC101715387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875064.3 | XP_004875121.1 | 2 - 3 | 646 | 73 | |
| XM_021252376.1 | XP_021108035.1 | 2 - 3 | 664 | 73 | ||
More Information
Set Membership: |
|

LOC101715387
Genomic Map