Assay Details
| Gene Symbol: Ccdc162p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068954.2 | XP_012924408.2 | 7 - 8 | 909 | 96 | |
| XM_021251515.1 | XP_021107174.1 | 7 - 8 | 909 | 96 | ||
| XM_021251516.1 | XP_021107175.1 | 7 - 8 | 909 | 96 | ||
| XM_021251517.1 | XP_021107176.1 | 7 - 8 | 909 | 96 | ||
| XM_021251518.1 | XP_021107177.1 | 7 - 8 | 909 | 96 | ||
| XM_021251519.1 | XP_021107178.1 | 6 - 7 | 795 | 96 | ||
| XM_021251520.1 | XP_021107179.1 | 7 - 8 | 909 | 96 | ||
| XM_021251521.1 | XP_021107180.1 | 7 - 8 | 909 | 96 | ||
More Information
Set Membership: |
|

Ccdc162p
Genomic Map