Assay Details
| Gene Symbol: LOC101717066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072797.2 | XP_012928251.1 | 5 - 6 | 1791 | 88 | |
| XM_021260072.1 | XP_021115731.1 | 4 - 5 | 703 | 88 | ||
More Information
Set Membership: |
|

LOC101717066
Genomic Map