Assay Details
| Gene Symbol: CUNH11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237718.1 | XP_021093377.1 | 6 - 7 | 730 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH11orf65| Gene Symbol: CUNH11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237718.1 | XP_021093377.1 | 6 - 7 | 730 | 108 | |
Set Membership: |
|