Assay Details
| Gene Symbol: LOC101716216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242809.1 | XP_021098468.1 | 11 - 12 | 1120 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101716216| Gene Symbol: LOC101716216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242809.1 | XP_021098468.1 | 11 - 12 | 1120 | 108 | |
Set Membership: |
|