Assay Details
| Gene Symbol: Necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256652.1 | XP_021112311.1 | 7 - 8 | 1201 | 59 | |
| XM_021256653.1 | XP_021112312.1 | 9 - 10 | 943 | 59 | ||
| XM_021256654.1 | XP_021112313.1 | 7 - 8 | 664 | 59 | ||
| XM_021256655.1 | XP_021112314.1 | 7 - 8 | 1087 | 59 | ||
| XM_021256656.1 | XP_021112315.1 | 7 - 8 | 601 | 59 | ||
| XM_021256657.1 | XP_021112316.1 | 7 - 8 | 1202 | 59 | ||
| XM_021256658.1 | XP_021112317.1 | 8 - 9 | 819 | 59 | ||
| XM_021256659.1 | XP_021112318.1 | 8 - 9 | 812 | 59 | ||
| XM_021256661.1 | XP_021112320.1 | 7 - 8 | 723 | 59 | ||
More Information
Set Membership: |
|

Necab2
Genomic Map