Assay Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844508.3 | XP_004844565.1 | 9 - 10 | 1163 | 60 | |
| XM_004844509.2 | XP_004844566.1 | 9 - 10 | 1154 | 60 | ||
| XM_004844510.3 | XP_004844567.1 | 8 - 9 | 955 | 60 | ||
| XM_021258154.1 | XP_021113813.1 | 9 - 10 | 1163 | 60 | ||
| XM_021258155.1 | XP_021113814.1 | 8 - 9 | 962 | 60 | ||
More Information
Set Membership: |
|

Lims1
Genomic Map