Assay Details
| Gene Symbol: Nfkbie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846448.3 | XP_004846505.1 | 4 - 5 | 1556 | 62 | |
| XM_004846450.3 | XP_004846507.1 | 5 - 6 | 1331 | 62 | ||
| XM_004846451.3 | XP_004846508.1 | 4 - 5 | 1544 | 62 | ||
More Information
Set Membership: |
|

Nfkbie
Genomic Map