Assay Details
| Gene Symbol: Pla2g2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851063.2 | XP_004851120.2 | 4 - 5 | 514 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g2e| Gene Symbol: Pla2g2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851063.2 | XP_004851120.2 | 4 - 5 | 514 | 113 | |
Set Membership: |
|