Assay Details
| Gene Symbol: Trmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864964.3 | XP_004865021.2 | 3 - 4 | 694 | 101 | |
| XM_013065735.2 | XP_012921189.1 | 3 - 4 | 696 | 101 | ||
| XM_021244537.1 | XP_021100196.1 | 3 - 4 | 512 | 101 | ||
| XM_021244538.1 | XP_021100197.1 | 3 - 4 | 527 | 101 | ||
More Information
Set Membership: |
|

Trmo
Genomic Map