Assay Details
Gene Symbol: Thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870219.2 | XP_004870276.1 | 7 - 8 | 1150 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870219.2 | XP_004870276.1 | 7 - 8 | 1150 | 63 |
Set Membership: |
![]() ![]() ![]() |