Assay Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871248.2 | XP_004871305.1 | 2 - 3 | 396 | 52 | |
| XM_021249446.1 | XP_021105105.1 | 2 - 3 | 399 | 52 | ||
More Information
Set Membership: |
|

Ntrk1
Genomic Map