Assay Details
| Gene Symbol: LOC101725179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874014.2 | XP_004874071.1 | 1 - 2 | 78 | 76 | |
| XM_013068981.2 | XP_012924435.1 | 1 - 2 | 78 | 76 | ||
| XM_021251572.1 | XP_021107231.1 | 1 - 2 | 78 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004874014.2, XM_013068981.2, XM_021251572.1 transcripts.
Set Membership: |
|

LOC101725179
Genomic Map