Assay Details
| Gene Symbol: LOC101725179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874014.2 | XP_004874071.1 | 3 - 4 | 385 | 101 | |
| XM_013068981.2 | XP_012924435.1 | 3 - 4 | 385 | 101 | ||
| XM_021251571.1 | XP_021107230.1 | 3 - 4 | 401 | 101 | ||
| XM_021251572.1 | XP_021107231.1 | 3 - 4 | 385 | 101 | ||
More Information
Set Membership: |
|

LOC101725179
Genomic Map