Assay Details
| Gene Symbol: LOC101725179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874014.2 | XP_004874071.1 | 4 - 5 | 505 | 81 | |
| XM_021251571.1 | XP_021107230.1 | 4 - 5 | 521 | 81 | ||
| XM_021251572.1 | XP_021107231.1 | 4 - 5 | 505 | 81 | ||
More Information
Set Membership: |
|

LOC101725179
Genomic Map