Assay Details
| Gene Symbol: LOC101725179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874014.2 | XP_004874071.1 | 5 - 6 | 682 | 71 | |
| XM_021251571.1 | XP_021107230.1 | 5 - 6 | 698 | 71 | ||
| XM_021251572.1 | XP_021107231.1 | 5 - 6 | 682 | 71 | ||
More Information
Set Membership: |
|

LOC101725179
Genomic Map